BR778013
BRAND® centrifuge tube, beaded rim
volume 15 mL, conical bottom, graduated: no
Synonym(s):
Centrifuge tubes, Tubes
All Photos(1)
About This Item
Recommended Products
material
borosilicate glass
conical bottom
feature
RCF 3,500 × g
graduated: no
packaging
pack of 100
manufacturer/tradename
BRAND 778013
volume
15 mL
Related Categories
General description
The Boro 3.3 Centrifuge Tube has a capacity of approximately 15 ml and features a robust wall thickness of 1.0 mm. Designed for high-performance applications, it can withstand relative centrifugal forces (RCF) of up to 3500. Ideal for laboratory use, this tube ensures durability and reliability during centrifugation, making it an essential tool for various scientific procedures.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG