Documents
DWK73802-15415
KIMBLE® solid cap with PTFE faced rubber liner
black phenolic cap, cap for 15-425
About This Item
UNSPSC Code:
41121800
NACRES:
NB.11
material
PTFE/rubber liner
black phenolic cap
sterility
non-sterile
feature
class A
packaging
case of 500 ea
manufacturer/tradename
Kimble® 73802-15415
fitting
cap for 15-425
General description
Class A Caps with PTFE-Faced Rubber Liners highly resistant to temperature and steam during autoclaving
  • Specially formulated phenolic cap material
  • Autoclavabl
  • White rubber liners
  • Excellent for general laboratory use
  • Identified in ASTM Specification E982, Class A requirement
Class A Caps with PTFE-Faced Rubber Liners highly resistant to temperature and steam during autoclaving
  • Specially formulated phenolic cap material
  • Autoclavable
  • White rubber liners
  • Excellent for general laboratory use
Identified in ASTM Specification E982, Class A requirement
Features and Benefits
• Specially formulated phenolic resin and liner adhesive to withstand the effects of repeated autoclaving• PTFE faced/general purpose white rubber liners are highly resistant to chemical effects• Excellent for general laboratory use• Identified in ASTM Specification E982, Class A requirements
Legal Information
KIMBLE is a registered trademark of DWK Life Sciences