Skip to Content
Merck
All Photos(1)

Key Documents

545686

Sigma-Aldrich

3-Chloro-2-methylbenzenesulfonyl chloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(CH3)SO2Cl
CAS Number:
Molecular Weight:
225.09
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

mp

71-74 °C (lit.)

SMILES string

Cc1c(Cl)cccc1S(Cl)(=O)=O

InChI

1S/C7H6Cl2O2S/c1-5-6(8)3-2-4-7(5)12(9,10)11/h2-4H,1H3

InChI key

ZSIYKAQPQRTBPF-UHFFFAOYSA-N

General description

3-Chloro-2-methylbenzenesulfonyl chloride is an aryl sulfonyl chloride derivative.

Application

3-Chloro-2-methylbenzenesulfonyl chloride may be used in the synthesis of ethyl [6-(3-chloro-2-methyl-benzenesulfonyl-amino)-pyridin-2-yl]-acetate and ethyl 2-(2-{[(3-chloro-2-methylphenyl)sulfonyl]amino}-1,3-thiazol-4-yl)acetate.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P G De Benedetti et al.
Journal of medicinal chemistry, 30(3), 459-464 (1987-03-01)
A set of 25 4'-, eight 2',4'-, and five 2',4',6'-substituted 4-aminodiphenyl sulfones were tested for their inhibitory activity on dihydropteroate synthase of Escherichia coli. Linear regression analysis shows that enzymic inhibition indices correlate well with both quantum chemical and spectroscopic
Tjeerd Barf et al.
Journal of medicinal chemistry, 45(18), 3813-3815 (2002-08-23)
Novel antidiabetic arylsulfonamidothiazoles are presented that exert action through selective inhibition of the 11beta-hydroxysteroid dehydrogenase type 1 (11beta-HSD1) enzyme, thereby attenuating hepatic gluconeogenesis. The diethylamide derivative 2a was shown to potently inhibit human 11beta-HSD1 (IC(50) = 52 nM), whereas the
Michael Siu et al.
Bioorganic & medicinal chemistry letters, 19(13), 3493-3497 (2009-05-29)
N-(Pyridin-2-yl) arylsulfonamides are identified as inhibitors of 11beta-hydroxysteroid dehydrogenase type 1 (11betaHSD1), an enzyme that catalyzes the reduction of the glucocorticoid cortisone to cortisol. Dysregulation of glucocorticoids has been implicated in the pathogenesis of diabetes and the metabolic syndrome. In

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service