Skip to Content
Merck
All Photos(1)

Documents

8.22049

Sigma-Aldrich

Dodecenylsuccinic anhydride

(mixture of isomers) for synthesis

Synonym(s):

Dodecenylsuccinic anhydride, 3-(2-Dodecylen-1-yl)-dihydro-2,5-furandione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H26O3
CAS Number:
Molecular Weight:
266.38
UNSPSC Code:
12352115
EC Index Number:
246-917-1
NACRES:
NA.22

vapor pressure

26 hPa ( 200 °C)

Quality Level

form

liquid

potency

2550 mg/kg LD50, oral (Rat)
6200 mg/kg LD50, skin (Rabbit)

pH

2-4 (20 °C, 50 g/L in H2O, slurry)

bp

200 °C/27 hPa

transition temp

flash point 127 °C

density

1.01 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C16H26O3/c1-2-3-4-5-6-7-8-9-10-11-12-14-13-15(17)19-16(14)18/h11-12,14H,2-10,13H2,1H3/b12-11+

InChI key

WVRNUXJQQFPNMN-VAWYXSNFSA-N

Analysis Note

Assay (morpholine method): ≥ 96.0 %
Density (d 20 °C/ 4 °C): 1.005 - 1.008
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

352.4 °F - closed cup

Flash Point(C)

178 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service