Skip to Content
Merck
All Photos(1)

Key Documents

47743

Sigma-Aldrich

4-Formylphenoxyacetic acid

≥97.0% (T)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H8O4
CAS Number:
Molecular Weight:
180.16
Beilstein:
779885
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

≥97.0% (T)

form

solid

mp

193-196 °C

storage temp.

2-8°C

SMILES string

OC(=O)COc1ccc(C=O)cc1

InChI

1S/C9H8O4/c10-5-7-1-3-8(4-2-7)13-6-9(11)12/h1-5H,6H2,(H,11,12)

InChI key

OYNIIKHNXNPSAG-UHFFFAOYSA-N

General description

4-Formylphenoxyacetic acid is a formylphenoxyaliphatic acid that reacts with dimedone to form 1,8-dioxo-octahydroxanthenes.[1] It participates in the synthesis of 4-substituted benzaldehyde derivatives[2] and 4-carboxypheoxyacetic acid.[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of 2-, 3-and 4-Carboxyphenoxyacetic Acid: A Necessary Modi?cation.
Adak MM.
Asian Journal of Chemistry, 5(2), 464-466 (1993)
Self-catalyzed syntheses, structural characterization, DPPH radical scavenging-, cytotoxicity-, and DFT studies of phenoxyaliphatic acids of 1, 8-dioxo-octahydroxanthene derivatives.
Kumar GSS, et al.
Journal of Molecular Structure, 1059, 51-60 (2014)
Wei Yi et al.
European journal of medicinal chemistry, 45(2), 639-646 (2009-11-26)
A series of novel 4-hydroxybenzaldehyde derivatives were synthesized and their inhibitory effects on the diphenolase activity of mushroom tyrosinase were investigated. Most of target compounds had more potent inhibitory activities than the parent compound 4-hydroxybenzaldehyde (IC(50)=1.22 mM). Interestingly, compound 3c

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service