Merck
  • Synthesis and biological evaluation of novel 4-hydroxybenzaldehyde derivatives as tyrosinase inhibitors.
Synthesis and biological evaluation of novel 4-hydroxybenzaldehyde derivatives as tyrosinase inhibitors.
European journal of medicinal chemistry (2009-11-26)
Wei Yi, Rihui Cao, Wenlie Peng, Huan Wen, Qin Yan, Binhua Zhou, Lin Ma, Huacan Song
ABSTRACT
A series of novel 4-hydroxybenzaldehyde derivatives were synthesized and their inhibitory effects on the diphenolase activity of mushroom tyrosinase were investigated. Most of target compounds had more potent inhibitory activities than the parent compound 4-hydroxybenzaldehyde (IC(50)=1.22 mM). Interestingly, compound 3c bearing a dimethoxyl phosphate was found to be the most potent inhibitor with IC(50) value of 0.059 mM. The inhibition kinetics analyzed by Lineweaver-Burk plots revealed that compound 3c was a non-competitive inhibitor (K(I)=0.0368 mM). In particular, compound 3c showed no side effects at dose of 1600 mg/kg in mice. These results suggested that such compounds might be served as lead compounds for further designing new potential tyrosinase inhibitors.
Sigma-Aldrich
4-Methoxycinnamic acid, predominantly trans, 99%
Sigma-Aldrich
4-Hydroxybenzaldehyde, ≥97%, FG
Sigma-Aldrich
4-Hydroxybenzaldehyde, ≥95.0% (HPLC)
Sigma-Aldrich
4-Hydroxybenzaldehyde, 98%