Documentos clave
SU860006
Supelco
Inter-Seal bonded caps (inseparable caps and liner)
white polypropylene cap (solid), tan PTFE/silicone (EPA Quality), thickness 3.2 mm, thread for 24-400, for use with 20 or 40mL vial, pkg of 100 ea
Seleccione un Tamaño
100 EA
111,00 €
111,00 €
Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad
Solicitar un pedido a granel
Seleccione un Tamaño
Cambiar Vistas
100 EA
111,00 €
About This Item
Código UNSPSC:
41122001
NACRES:
NB.11
111,00 €
Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad
Solicitar un pedido a granel
Materiales
tan PTFE/silicone (EPA Quality)
white polypropylene cap (solid)
Características
InterSeal (cap and liner form an unseparatable unit)
dureza
45 (Durometer: shore A)
envase
pkg of 100 ea
grosor
3.2 mm
Adaptador
thread for 24-400
compatibilidad
for use with 20 or 40mL vial
Descripción general
Inter-Seal bonded caps eliminate expensive and time consuming hand assembly while preventing liner fallout. They are made by a bonding process that bonds PTFE/silicone into polypropylene closures without using adhesives. The bonding properly seats and secures the septa within the vial opening. This integrated one piece system withstands multiple injections, making it very cost effective.
Benefits include:
  • Significant bond strength
  • Withstands multiple injections
  • Resistant to coring
  • Excellent resealability
  • Broad chemical resistance
Información legal
Inter-Seal is a trademark of Integrated Liner Technologies, Inc.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)