Documentos clave
57476
Needle for Model 3725i Prep Sample Injector
stainless steel, needle size 16 ga
About This Item
Código UNSPSC:
41115854
Materiales
stainless steel
tamaño de la aguja
16 ga
envase
pkg of 1
compatibilidad
for use with Rheodyne 3725-086
Descripción general
Needle for Model 3725i Prep Sample Injector includes a 5 cm large-bore syringe needle (16 gauge/0.65 in. OD) for rapid injection of large samples.
Productos recomendados
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)