Merck
51384-U
EXP® Fitting System
hand tight nut and titanium hybrid ferrule, pack of 1
About This Item
Código UNSPSC:
41115700
NACRES:
SB.52
envase
pack of 1
Descripción general
The EXP® Fitting System is the premier adjustable nut and ferrule compression fitting for extreme high-pressure connections between 1/16 inch tubing and any 10-32 port. The Titanium Hybrid ferrule provides a perfect seal with every connection, yet can be released without tools to adjust to the different port depths of various hardware. With this fitting system there is no longer the need to clip off and replace ferrules from tubing.
  • Auto-adjusting Zero Dead Volume (ZDV) connection
  • Intended for many repeat uses
  • Rated to 20,000 psi
Productos recomendados
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Información legal
EXP is a registered trademark of Optimize Technologies, Inc.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)