Documentos clave
33195
Supelco
Crimper
Hand crimper, for use with 11mm crimp seals, pkg of 1 ea
Seleccione un Tamaño
1 EA
437,00 €
437,00 €
Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad
Solicitar un pedido a granel
Seleccione un Tamaño
Cambiar Vistas
1 EA
437,00 €
About This Item
Código UNSPSC:
41122001
437,00 €
Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad
Solicitar un pedido a granel
envase
pkg of 1 ea
compatibilidad
for use with 11mm crimp seals
Descripción general
The Supelco hand held crimper provides a consistent and dependable seal that allows secure vial closure each and evey time. The high quality construction is engineered for durability and long life to provide a smooth and simple operation.
Aplicación
Hand crimper may be used to seal a glass vial containing MeOH, for high-performance liquid chromatography-diode array detection (HPLC-DAD) analysis, during polyurethanes (PUs) analysis and clean-up, in an experimental study done in order to develop, optimise and apply PU as new polymeric phases for stir bar sorptive extraction.[1]
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)
Visite la Librería de documentos
N R Neng et al.
Journal of chromatography. A, 1171(1-2), 8-14 (2007-10-12)
In this contribution, polyurethane foams are proposed as new polymeric phases for stir bar sorptive extraction (SBSE). Assays performed for polyurethane synthesis demonstrated that four series of formulations (P(1), P(2), P(3) and P(4)) present remarkable stability and excellent mechanical resistance