Documentos clave
Z757756
Sartorius pipette tips
volume range 50-1200 μL, standard, refill, sterile
Seleccione un Tamaño
960 EA
87,88 €
87,88 €
Precio de catálogo94,50 €Ahorre 7 %
Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad
Seleccione un Tamaño
Cambiar Vistas
960 EA
87,88 €
About This Item
Código UNSPSC:
41121600
NACRES:
NB.01
87,88 €
Precio de catálogo94,50 €Ahorre 7 %
Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad
Materiales
colorless
esterilidad
sterile
Características
DNase free
RNase free
barrier: no
endotoxin free
envase
pack of 96 ea (Racked)
fabricante / nombre comercial
Sartorius 791203
volumen máximo
1.2 mL
intervalo de volumen
50-1200 μL
volumen
50—1250 μL
Categorías relacionadas
Descripción general
The non-filtered Optifit Tips are designed for best possible fitting and sealing to secure the highest possible accuracy and precision. These pipette tips enable ergonomic and light tip attachment.
Optifit Tips are available in various packaging types for all your needs:
  • Single Tray racks – purity certified tip racks
  • Refill Packs - purity certified and ecological refill packages
  • Refill Towers - space-saving and ecological refill packages
  • Flexibulk boxes - space-saving, ecological and economical bulk boxes
  • Standard bulk – economical bulk boxes
While Optifit Tip trays are colour coded for easy matching with a corresponding, colour coded Sartorius pipette, they do also match with most other pipette due to their universal design.
Strict quality standards, ISO 9001 and ISO 13485, are followed from tip R&D to production and delivery. The purity certified tips are manufactured in ISO 8 classified clean room conditions to avoid any contaminations and are tested for DNase, RNase and endotoxins per each tip lot. Pre-sterilised tips are also available for the purest laboratory work. Sartorius offers a selection of pipette tips, extended and large volume tips, for special needs.
Otras notas
Browse all pipette tips
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)