Documentos clave
Z740376
Eco-Friendly SpecTainer, tamper evident, biodegradable
size 60 mL, non-sterile
Seleccione un Tamaño
500 EA
563,00 €
563,00 €
Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad
Seleccione un Tamaño
Cambiar Vistas
500 EA
563,00 €
About This Item
Código UNSPSC:
41121800
NACRES:
NB.17
563,00 €
Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad
Materiales
polyethylene cap
polypropylene container
esterilidad
non-sterile
envase
pack of 500 ea
fabricante / nombre comercial
Simport C566-60DOECO
tamaño
60 mL
Descripción general
The Simport Eco-Friendly SpecTainer will biodegrade to become some of the soil′s organic components in less than 7 years in constant contact with the soil instead of up to 400 years when using conventional plastics.
The Simport Eco-Friendly SpecTainer, one of the most innovative specimen containers on the market today, is now available in a new version: The Eco-Friendly SpecTainer, a rapidly degradable Specimen Container. Plastics take hundreds of years to degrade naturally in the environment. The only condition that is necessary for the Eco-Friendly SpecTainer to biodegrade is constant contact with other degrading material. No heat, physical stress, oxygen or sunlight necessary. The Eco-Friendly SpecTainer has the ability to break down safely and quickly, by biological means, into the raw materials of nature and disappear into the environment.
Does not contain heavy metals. No negative effect to its physical properties. No heat, light or mechanical stress needed for product breakdown. No toxic residue. Fully degradable within 7 years, aerobically or anaerobically.
Información legal
SpecTainer is a trademark of Simport
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)