Saltar a
Z688339
LightSafe centrifuge tubes
capacity 50 mL, conical polypropylene, sterile
Iniciar sesión para ver los precios por organización y contrato.
Acerca de este artículo
NACRES:
NB.22
UNSPSC Code:
41121703
eCl@ss:
32040302
Capacity:
50 mL
Material:
black tube, cap (attached), conical polypropylene
Sterility:
sterile
Packaging:
pack of 50
Feature:
RCF 10,000 × g
material
black tube, cap (attached), conical polypropylene
sterility
sterile
feature
RCF 10,000 × g
packaging
pack of 50
manufacturer/tradename
Cole Parmer 634435
capacity
50 mL
General description
Disposable polypropylene tubes and caps with recessed lid to prevent leaking. Frosted writing surface allows for permanent identification and labeling. Autoclavable to 122°C and can be stored in freezers to -100°C. RCF values: 15 mL, 8,400 g/50 mL, 10,000 g
Still not finding the right product?
Explore all of our products under LightSafe centrifuge tubes
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)