T0567
Gel staining tray
L × W 7 1/2 in. (19 cm) × 7 1/2 in. (19 cm)
About This Item
Código UNSPSC:
41105300
NACRES:
NB.51
Productos recomendados
L × An
7 1/2 in. (19 cm) × 7 1/2 in. (19 cm)
Descripción general
Optically clear down to 300 nm; photograph gels with UV or visible light right in the tray. Small feet at corners protect bottom from scratching. Stackable: rock several gels at once; stacking seals tray below to prevent evaporation and spills. For use with aqueous staining and destaining solutions only.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)