Seleccione un Tamaño
| Talla/SKU | Disponibilidad | Precio |
|---|---|---|
1 ea | Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad | 26,30 € |
Acerca de este artículo
26,30 €
material
high-density polypropylene (coated steel wire), white holder
feature
holds 1 x Glove box
packaging
pack of 1 ea
manufacturer/tradename
Heathrow Scientific HS23457
L × W × H
144 mm × 209 mm × 109 mm
General description
Features and Benefits
- One-Box Holder
- HDPE coated steel wire racks are an economical solution
- Free up bench-top space by mounting holders on wall for easy dispensing of gloves
- Universal fit for most major brands of glove boxes
- Mounting screws included
1 of 1
Este artículo | |||
|---|---|---|---|
| description white holder | description Holds 3 boxes, epoxy-coated (steel wire) | description anti-microbial, W × L × H 254 mm × 156 mm × 97 mm, blue | description W × L × H 254 mm × 156 mm × 97 mm, white |
| manufacturer/tradename Heathrow Scientific HS23457 | manufacturer/tradename Heathrow Scientific HS23458 | manufacturer/tradename Heathrow Scientific HS23491A | manufacturer/tradename Heathrow Scientific HS120258 |
| feature holds 1 x Glove box | feature Holds 3 boxes | feature Holds 1 box of gloves | feature Holds 1 box |
| material high-density polypropylene (coated steel wire), white holder | material epoxy-coated (steel wire) | material plastic | material plastic |
| packaging pack of 1 ea | packaging - | packaging - | packaging - |
| L × W × H 144 mm × 209 mm × 109 mm | L × W × H - | L × W × H - | L × W × H - |
Elija entre una de las versiones más recientes:
¿Ya tiene este producto?