Seleccione un Tamaño
| Talla/SKU | Disponibilidad | Precio |
|---|---|---|
1 ea | Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad | 22,80 € |
Acerca de este artículo
22,80 €
material
blue , nickel closure
feature
Holds 100 x slides 76x26 mm
manufacturer/tradename
Heathrow Scientific HS15994A
L × W × H
208 × 175 × 34 mm
General description
Features and Benefits
- molded, rigid boxes securely hold slides in grooved slots
- durable ABS plastic boxes are available wth either cork or foam lining
- both the lining and the slots are numbered to correspond to the slide inventory sheet
- closes securely with a rust-resistant, nickel-plated clasp and pin hinges
1 of 1
Este artículo | |||
|---|---|---|---|
| description 100 Place, cork lining, blue | description 100 Place, foam lining, white | description 100 Place, cork lining, black | description 25 Place, cork lining, blue |
| feature Holds 100 x slides 76x26 mm | feature Holds 100 x slides 76x26 mm | feature Holds 100 x slides 76x26 mm | feature Holds 25 x slides 76x26 mm |
| manufacturer/tradename Heathrow Scientific HS15994A | manufacturer/tradename Heathrow Scientific HS15994R | manufacturer/tradename Heathrow Scientific HS15994G | manufacturer/tradename Heathrow Scientific HS15989A |
| material blue , nickel closure | material white , nickel closure | material nickel closure, black | material blue , nickel closure |
| L × W × H 208 × 175 × 34 mm | L × W × H 208 × 175 × 34 mm | L × W × H 208 × 175 × 34 mm | L × W × H 208 × 175 × 34 mm |
Still not finding the right product?
Explore all of our products under Premium Slide box
Elija entre una de las versiones más recientes:
¿Ya tiene este producto?