HS120248
Wide mouth wash bottle
capacity 500 mL, green PE-LD bottle
Seleccione un Tamaño
35,40 €
Seleccione un Tamaño
About This Item
35,40 €
Productos recomendados
Materiales
green PE-LD bottle
polypropylene cap
Características
label no label
vented
with cap
envase
pack of 6 ea
fabricante / nombre comercial
Heathrow Scientific HS120248
capacidad
500 mL
Categorías relacionadas
Descripción general
The closure and stem are molded in one piece to make them leak-proof. The angled neck and tip provide a uniform stream of liquid and the tip can be cut back if required to increase the flow.
- Wide bottle mouth makes filling easier
- Color-coding for user, application or location identification
- Bottle is made from LDPE, with a PP closure
- 500 mL bottle size,
- Supplied in packs of 6
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)