Seleccione un Tamaño
| Talla/SKU | Disponibilidad | Precio |
|---|
Acerca de este artículo
material
white polystyrene (Anti-static)
packaging
pk of 500
manufacturer/tradename
Heathrow Scientific HS120225
L × W × H
43 mm × 58 mm × 13 mm
General description
Features and Benefits
- biologically inert and resistant to dilute and weak acids, aqueous solutions, alcohols, and bases.
- temperature range -10 to 70C
- disposable
- pour-spout design easily dispenses samples without flexing the dish
- flat-bottom dishes are made from 12 mil think polystyrene for better strength, stability, and handling
1 of 1
Este artículo | |||
|---|---|---|---|
| description small, white polystyrene (Anti-static), pk of 500 | description small, white polystyrene, pack of 500 ea | description large, white polystyrene (Anti-static), pk of 500 | description medium, white polystyrene (Anti-static), pk of 500 |
| manufacturer/tradename Heathrow Scientific HS120225 | manufacturer/tradename Heathrow Scientific HS1419A | manufacturer/tradename Heathrow Scientific HS120227 | manufacturer/tradename Heathrow Scientific HS120226 |
| material white polystyrene (Anti-static) | material white polystyrene | material white polystyrene (Anti-static) | material white polystyrene (Anti-static) |
| packaging pk of 500 | packaging pack of 500 ea | packaging pk of 500 | packaging pk of 500 |
| L × W × H 43 mm × 58 mm × 13 mm | L × W × H 43 mm × 58 mm × 13 mm | L × W × H 108 mm × 183 mm × 26 mm | L × W × H 83 mm × 132 mm × 26 mm |
Still not finding the right product?
Explore all of our products under Pour Boat Weighing Dishes
Elija entre una de las versiones más recientes:
¿Ya tiene este producto?