Saltar a
HS120218
Pearl™ Pipette
adjustable, volume 30-300 μL, blue, 12-channel
Iniciar sesión para ver los precios por organización y contrato.
Seleccione un Tamaño
Cambiar Vistas
Talla/SKUDisponibilidadPrecio
1 ea
Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad
714,00 €
Acerca de este artículo
NACRES:
NB.01
UNSPSC Code:
41121500
Material:
blue , white
714,00 €
Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad
​
Servicio técnico
¿Necesita ayuda? Nuestro equipo de científicos experimentados está aquí para ayudarle.
Permítanos ayudarle
material
blue , white
feature
12-channel, adjustable
packaging
pack of 1 ea
manufacturer/tradename
Heathrow Scientific HS120218
volume measuring range
30-300 μL
General description
The Pearl is a single-channel pipette offering adjustable volume and precision liquid handling. The sleek, ergonomic design and reduced plunger pressure help reduce the chance of Repetitive Stress Injury (RSI), as the pipette requires 2/3 the operation pressure of most standard pipettes. The Pearl is tested in accordance with standards and a calibration certificate is supplied. Manufactured in accordance with ISO08655 and GLP regulations.
Features and Benefits
  • Internal metal parts for increased accuracy and calibration stability, and long life
  • Ergonomic Slight downward slant to tip ejector button reduces hand fatigue, incidence of repetitive strain injury (RSI)
  • Easy volume adjustment with locking tab to prevent accidental change
  • Separate tip ejector
  • Large digit volume display
  • Color coded and labeled with volume range
  • Both top and side have colored coded components for quick visual confirmation
  • CE certified and Manufactured in accordance with ISO08655 and GLP regulations
  • Autoclavable
Legal Information
Pearl is a trademark of Specac, Inc.
Comparar elementos similares
Ver comparación completa
Mostrar Diferencias
1 of 1
Este artículo
HS120101HS120097HS120216
description
adjustable, volume 30-300 μL, blue, 12-channel
description
adjustable, volume 10-100 μL, blue, single-channel
description
adjustable, volume 0.1-2 μL, green, single-channel
description
adjustable, volume 30-300 μL, purple, 8-channel
material
white , blue
material
white , blue
material
white , green
material
white , purple
feature
12-channel, adjustable
feature
adjustable, single-channel
feature
single-channel, adjustable
feature
adjustable, 8-channel
volume measuring range
30-300 μL
volume measuring range
10-100 μL
volume measuring range
0.1-2 μL
volume measuring range
30-300 μL
manufacturer/tradename
Heathrow Scientific HS120218
manufacturer/tradename
Heathrow Scientific HS120101
manufacturer/tradename
Heathrow Scientific HS120097
manufacturer/tradename
Heathrow Scientific HS120216
packaging
pack of 1 ea
packaging
pack of 1 ea
packaging
pack of 1 ea
packaging
pack of 1 ea
Still not finding the right product?
Explore all of our products under Pearl™ Pipette
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)