Documentos clave
GE18-1153-14
Sigma-Aldrich
TRICORN 10/50 GLASS COLUMN
Cytiva 18-1153-14, pack of 1 ea
Seleccione un Tamaño
1 EA
399,00 €
399,00 €
Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad
Seleccione un Tamaño
Cambiar Vistas
1 EA
399,00 €
About This Item
Código UNSPSC:
41121800
NACRES:
SB.52
399,00 €
Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad
Materiales
glass column
Características
wetted part
envase
pack of 1 ea
fabricante / nombre comercial
Cytiva 18-1153-14
Categorías relacionadas
Descripción general
Glass tube 10/50
Características y beneficios
  • Tricorn columns are designed for high resolution liquid chromatography of biomolecules performed at medium to high pressures (see Specifications). They are suitable for use with high performance media, such as Sepharose High Performance, Superdex Prep Grade, and SOURCE.
  • The columns are available with inner diameters of 5 and 10 mm, and in lengths from 20 to 200 mm. Column lengths of 300 and 600 mm are also available for the 10 mm diameter columns. The design and dimensions of the columns make them particularly useful for screening and polishing steps in protein purification.
  • Glass tubes coated with plastic film protects individuals in the laboratory from harm if the column is dropped or operated outside its specification.
Productos recomendados
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)