GE18-1153-07
Tricorn Glass Tube
joint: 5/150
Seleccione un Tamaño
408,00 €
Seleccione un Tamaño
About This Item
408,00 €
Junta
joint: 5/150
fabricante / nombre comercial
Cytiva 18-1153-07
18-1153-13
Descripción general
- High performance glass columns designed for reliable high resolution, reproducibility and durability
- New longer adaptor for increased adjustment of bed height
- Modern, integrated design – simple to assemble, pack, operate, and maintain
- Broad range of column dimensions – suitable for many different purification needs
- Safety first – glass tubes coated with plastic film to protect laboratory personnel if the column is dropped or operated outside its specifications
- Locking ring – prevents accidental compression of media bed
- Separate packing connector – allows simple column packing for a broad range of media
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)