EP4925000049
Eppendorf® Reference® 2 G
single-channel, fixed, volume (10 μL), medium gray
Seleccione un Tamaño
218,00 €
Seleccione un Tamaño
About This Item
218,00 €
Productos recomendados
Características
single-channel, fixed
envase
pack of 1 ea
fabricante / nombre comercial
Eppendorf® 4925000049
volumen
(10 μL)
color
medium gray
Categorías relacionadas
Descripción general
Besides improved ergonomics the design focus lays on providing the most accurate results possible, optimum user safety, reliability and robustness for a long service life. The innovative single button operation is not only fast and easy, but it also allows active aerosol1) reduction and thus protects the user, the sample and the pipette. The sturdy handle manufactured with stainless steel protects the pipette from impact and in combination with the spring loaded bumpers the Reference 2 is likely to stay within calibration even if it takes a fall. These high levels of security make the Reference 2 ideal for use with precious liquids, or in any application requiring extremely high levels of accuracy. When there can be no compromise on quality and reproducibility, Eppendorf′s Reference 2 is an ideal instrument for these liquid handling applications.
Single-Button Operation
- Lower Operating forces for aspiration/dispensing/tip ejection limit strain on the user.
- Allows users to move more quickly through their pipetting tasks by not having to move the thumb from side to side to eject tips.
- With the ability to eject a tip immediately following dispensing, aerosol contamination of your pipette is considerably reduced.
Información legal
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)