Documentos clave
BSIPSL3ANC
BS/IP/SL Viscometer for Transparent Liquids
Size 3A Nom. Constant 3.0, Range 600 to 3,000 cSt, (non certified)
About This Item
Código UNSPSC:
41116107
NACRES:
NA.24
fabricante / nombre comercial
Paragon Scientific Ltd
aplicaciones
cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care
Descripción general
This is the non certified version of this viscometer. If you wish to order a certifed viscometer, please refer to the corresponding product BSIPSL3AUKC
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)