Documentos clave
XX4304704
Millipore
Hose adapter, 1/4 in. NPTF to 5/16 in. ID, polypropylene
Accessories for filter holders for sample preparation.
Seleccione un Tamaño
2 EA
56,60 €
56,60 €
Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad
Seleccione un Tamaño
Cambiar Vistas
2 EA
56,60 €
About This Item
Código UNSPSC:
41104909
eCl@ss:
32031613
NACRES:
NB.24
56,60 €
Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad
Materiales
polypropylene adapter
Nivel de calidad
D.I.
6.3 mm (0.25 in.)
Condiciones de envío
ambient
Descripción general
Millipore has a broad selection of fittings, adapters, valves, gauges, and tubing that can be used in conjunction with Millipore filtration systems. With stated exceptions, material of construction is Series 300 stainless steel.
Tri-Clamp (TC) fittings are referred to by convention as 1 1/2", 2", etc. 1 1/2" TC fittings connect to 1" or 1 1/2" O.D. tube sizes. 2" TC fittings connect to 2" O.D. tube size only.