Documentos clave
EZSSGASK1
Millipore
Silicon gasket for EZ-Stream® pump
To replace original gasket
Seleccione un Tamaño
1 EACH
25,00 €
25,00 €
Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad
Solicitar más información
Seleccione un Tamaño
Cambiar Vistas
1 EACH
25,00 €
About This Item
Código UNSPSC:
41116500
NACRES:
NB.79
25,00 €
Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad
Solicitar más información
Características
repair kit
envase
pkg of 1 numbers
fabricante / nombre comercial
EZ-Stream®
aplicaciones
bioburden testing
food and beverages
industrial qc
pharmaceutical
water monitoring
compatibilidad
for use with EZ-Stream®
Descripción general
The silicon gasket is for the replacement of the original gasket that ensures the tightness between the housing and the bottom plate of the EZ-Stream® vacuum pump to prevent liquid to seep inside the pump. The EZ-Stream® vacuum pump is a liquid transfer pump with a compact design that makes the pump ideal for use on both the workbench and in the laminar flow hood
Aplicación
The silicon gasket ensures tightness between the housing and the bottom plate of the EZ-Stream® pump that is used for microbiology analysis of Beer, Dairy, Bottled Water, Pharmaceutical Products Analysis, Soft Drink, Cider, Sports Drink & Wine.
Información legal
EZ-STREAM is a registered trademark of Merck KGaA, Darmstadt, Germany