Saltar al contenido
Merck
Todas las fotos(2)

Documentos clave

ABN338

Sigma-Aldrich

Anti-Slitrk4 Antibody

from rabbit, purified by affinity chromatography

Sinónimos:

SLIT and NTRK-like protein 4, Slitrk4

Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización

Seleccione un Tamaño

100 μG
390,00 €

390,00 €


Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad

Solicitar un pedido a granel

Seleccione un Tamaño

Cambiar Vistas
100 μG
390,00 €

About This Item

Código UNSPSC:
12352203
eCl@ss:
32160702
NACRES:
NA.41

390,00 €


Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad

Solicitar un pedido a granel

origen biológico

rabbit

Nivel de calidad

forma del anticuerpo

affinity isolated antibody

tipo de anticuerpo

primary antibodies

clon

polyclonal

purificado por

affinity chromatography

reactividad de especies

mouse, human

técnicas

immunohistochemistry: suitable
western blot: suitable

Nº de acceso UniProt

Condiciones de envío

wet ice

modificación del objetivo postraduccional

unmodified

Información sobre el gen

Descripción general

SLIT and NTRK-like family 4 (Slitrk4) is a member a protein family consisting of six homologous transmembrane proteins (Slitrk1-6) that share two conserved leucine-rich repeat domains in the extracellular domain and have significant homology to Slit, a secreted axonal growth-controlling protein. These proteins are also homologous to trk neurotrophin receptors in their intracellular domains. Expression of Slitrk proteins is highly restricted to neural and brain tumor tissues, but varies within the protein family. Like every other Slitrk protein except Slitrk1, overexpression of Slitrk4 inhibited neurite outgrowth in cultured neurons, suggesting that these proteins are involved in the control of neurite outgrowth.

Especificidad

This antibody recognizes the C-terminus of Slitrk4.

Inmunógeno

KLH-conjugated linear peptide corresponding to human Slitrk4.

Aplicación

Immunohistochemistry Analysis: 2.5 μg/mL from a representative lot detected Slitrk4 in human brain tissue.
Research Category
Neuroscience
Research Sub Category
Developmental Neuroscience
This Slitrk4 antibody is validated for use in WB & IHC for the detection of the Slitrk4 protein.

Calidad

Evaluated by Western Blot in mouse brain tissue lysate.

Western Blot Analysis: 1 µg/mL of this antibody detected Slitrk4 in 15 µg of mouse brain tissue lysate.

Descripción de destino

~94 kDa observed

Forma física

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing PBS with 0.02% sodium azide.

Almacenamiento y estabilidad

Stable for 1 year at 2-8°C from date of receipt.

Nota de análisis

Control
Mouse brain tissue lysate

Otras notas

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Cláusula de descargo de responsabilidad

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

¿No encuentra el producto adecuado?  

Pruebe nuestro Herramienta de selección de productos.

Código de clase de almacenamiento

10 - Combustible liquids

Clase de riesgo para el agua (WGK)

WGK 1


Certificados de análisis (COA)

Busque Certificados de análisis (COA) introduciendo el número de lote del producto. Los números de lote se encuentran en la etiqueta del producto después de las palabras «Lot» o «Batch»

¿Ya tiene este producto?

Encuentre la documentación para los productos que ha comprado recientemente en la Biblioteca de documentos.

Visite la Librería de documentos

Preguntas

Revisiones

Sin puntuación

Filtros activos

Nuestro equipo de científicos tiene experiencia en todas las áreas de investigación: Ciencias de la vida, Ciencia de los materiales, Síntesis química, Cromatografía, Analítica y muchas otras.

Póngase en contacto con el Servicio técnico