Documentos clave
Z743133
Julabo® cryo-compact circulator
CF30, capacity 3.5 L, temperature -30-150 °C, precision: ±0.03, AC/DC input 230 V AC, UK plug
Seleccione un Tamaño
1 EA
5530,00 €
5530,00 €
Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad
Seleccione un Tamaño
Cambiar Vistas
1 EA
5530,00 €
About This Item
Código UNSPSC:
41103706
NACRES:
NB.77
5530,00 €
Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad
envase
pack of 1 ea
fabricante / nombre comercial
Julabo 9400330
intervalo de medición de la temperatura
-30-150 °C, precision: ±0.03
capacidad
3.5 L
Descripción general
The new CF series offers compact refrigerated/heating circulators. With small overall dimensions the units can even be placed on a very small surface or within a complex installation. All models offer a heating capacity of 2 kW and safety class 3 according to DIN12876-1. The maximum ambient temperature of +40°C as well as a ventilation air cooling allow installing the units very closely side by side with other units or directly under a fume hood. Furthermore all the models feature pump connections for external temperature applications and a bath opening for temperature control of small objects directly in the circulator bath. Wetted parts are made of high quality stainless steel or plastic. The instruments provide a reliable Microprocessor electronics with high temperature stability as well as warning and safety functions.
Información legal
Julabo is a registered trademark of Julabo USA, Inc.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)