Documentos clave
Z607614
Bohlender HT (high temp) laboratory screw joints
GL 14 thread, tubing O.D. 8.0 mm
Seleccione un Tamaño
1 EA
33,60 €
33,60 €
Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad
Seleccione un Tamaño
Cambiar Vistas
1 EA
33,60 €
About This Item
Código UNSPSC:
41121815
NACRES:
NB.35
33,60 €
Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad
fabricante / nombre comercial
Bohlender
disponibilidad
not available in USA and Canada
tuberías D.E.
8.0 mm
Adaptador
GL 14 thread
Descripción general
Pressure resistant up to approx. 10 bar (at room temperature for thread sizes GL 14, GL 18 and GL 25), suitable for vacuum. The circulating medium is exposed only to PTFE; screw cap made of glass-fibre PPS (max.+250 °C). For many years laboratory screw joints have been mainly used for connecting glass tubes with PTFE tubing or with plastic, glass and metal tubes. In addition, these couplings are also suitable for locking probes (even stainless steel), thermometers, laboratory stopcocks, immersion tubes or lead-in cables.
Información legal
Bohlender is a trademark of Bohlender GmbH
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)