This product is a standard Combi-Clip that fits a 24/40 joint, as shown below.
Z566616
Buchi Combi-Clip
fits 24/40 joint
Seleccione un Tamaño
42,60 €
Seleccione un Tamaño
About This Item
42,60 €
Productos recomendados
envase
pack of 1 ea
fabricante / nombre comercial
Büchi 040620
Descripción general
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)
Los clientes también vieron
-
Does this clip fit the thread on the RE-111?
1 respuesta-
¿Le ha resultado útil?
-
-
Ist der Combi-Clip kompatibel mit einem Büchi Rotavapor R-300?
1 respuesta-
Yes, this combi-clip is compatible with the Büchi Rotavapor R-300.
¿Le ha resultado útil?
-
-
What is the Department of Transportation shipping information for this product?
1 respuesta-
Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.
¿Le ha resultado útil?
-
-
For the Buchi Combi-Clip, Product Z566616, what Buchi model rotary evaporator (rotovap) does it fit?
1 respuesta-
This Combi-Clip fits only Buchi models R-200/205 and R-210/215. It is usually used with a vapor duct that looks like the one in the attached photo. It will not fit some older Buchi models that have a more narrow duct. The ducts for Buchi models R114/124 (Z403245, short and Z403237, long) already come with a Combi-Clip, so if one of those ducts is ordered, there is no need to order a separate Combi-Clip.
¿Le ha resultado útil?