Z555592
Aldrich® two-neck solvent storage flask
capacity 500 mL
About This Item
Productos recomendados
Materiales
borosilicate glass
round bottom flask
two-neck
esterilidad
non-sterile
Características
closure type Standard Taper Joint
envase
pack of 1
fabricante / nombre comercial
Sigma-Aldrich
capacidad
500 mL
Descripción general
PTFE stopcock, ST 24/40 joints.
Información legal
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC