Merck
Z543853
Sigma-Aldrich
Compression tubing fitting for VerSA-Flow dispenser
tubing O.D. 1/8 in.
About This Item
Código UNSPSC:
41105108
tuberías D.E.
1/8 in.
Descripción general
Adapts rigid wall tubing of various sizes to VerSA-Flow caps. Fitting includes nut with ferrules and PBT bushing with 1/4 in. NPTM threads.
Información legal
VerSA-Flow is a trademark of Sigma-Aldrich Co. LLC
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)