Documentos
Z302848
Quickfit® round-bottom flasks
capacity 500 mL, joint: ST/NS 29/32
Sinónimos:
FR500/4S
About This Item
Código UNSPSC:
41121804
NACRES:
NB.43
Materiales
borosilicate glass
round bottom flask
short
single-neck
Junta
joint: ST/NS29/32
esterilidad
non-sterile
Características
closure type Standard Taper Joint
envase
pack of 1
case of 5
fabricante / nombre comercial
Quickfit FR500/4S
disponibilidad
not available in USA and Canada
capacidad
500 mL
Descripción general
Short neck with ground-glass joint.
Información legal
Quickfit is a registered trademark of SciLabware Limited