Documentos clave
Z280178
Sigma-Aldrich
Tygon® formula R-3603 laboratory tubing
I.D. × O.D. 1/4 in. × 5/8 in.
About This Item
Código UNSPSC:
41100000
envase
pkg of 10 feet
fabricante / nombre comercial
Tygon
D.I. × D.E.
1/4 in. × 5/8 in.
pared tamaño
3/16 in.
Descripción general
Clear, general-purpose laboratory tubing. Ideal for peristaltic pumps, autoclavable to 248°F for 30 min. Temp. range: -49 to 158°F. Heavy walled Tygon is suitable for vacuum use.
  • Resists most inorganic chemicals
  • Resistant to cracking and aging
  • Low vapor pressure
  • Lasts longer than rubber tubing
Aplicación
suitable for vacuum
Información legal
Tygon is a registered trademark of Saint-Gobain Performance Plastics Corp.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)