Seleccione un Tamaño
| Talla/SKU | Disponibilidad | Precio |
|---|---|---|
1 ea | Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad | 27,10 € |
Acerca de este artículo
27,10 €
material
borosilicate glass flask, pear-shaped , single-neck
joint
joint: ST/NS14/10
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F321010
capacity
10 mL, 10 mL
General description
Legal Information
1 of 1
Este artículo | |||
|---|---|---|---|
| description 10 mL, joint: ST/NS 14/10 | description 5 mL, joint: ST/NS 14/10 | description 10 mL, joint: ST/NS 14/20 | description 25 mL, joint: ST/NS 14/10 |
| manufacturer/tradename Synthware F321010 | manufacturer/tradename Synthware F321005 | manufacturer/tradename Synthware F321410L | manufacturer/tradename Synthware F321025 |
| capacity 10 mL, 10 mL | capacity 5 mL , 5 mL, 50 mL | capacity 10 mL , 10 mL | capacity 25 mL, 25 mL |
| joint joint: ST/NS14/10 | joint joint: ST/NS14/10 | joint joint: ST/NS14/20 | joint joint: ST/NS14/10 |
| material borosilicate glass flask, pear-shaped , single-neck | material pear-shaped , single-neck , borosilicate glass flask | material borosilicate glass flask, single-neck , pear-shaped | material borosilicate glass flask, single-neck , pear-shaped |
| packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea |
Still not finding the right product?
Explore all of our products under Synthware™ pear shaped flask
Elija entre una de las versiones más recientes:
¿Ya tiene este producto?