Documentos clave
SLW5810/3
Pyrex® Vacuum filtration system for 47 mm membranes
support base only
Sinónimos:
Pyrex, SciLabware 5810/3
Seleccione un Tamaño
1 EA
350,00 €
350,00 €
Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad
Seleccione un Tamaño
Cambiar Vistas
1 EA
350,00 €
About This Item
Código UNSPSC:
41104925
NACRES:
NB.43
350,00 €
Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad
envase
pkg of 1 ea
fabricante / nombre comercial
5810/3
SciLabware
Descripción general
This all glass vacuum filtration assembly, comprising of a 300 ml capacity graduated funnel, support base with integral sintered glass filter disc, 1000 ml conical flask and metal retaining clamp, is ideal for the preparation of HPLC solvents or for microbiology filtration applications.
Características y beneficios
  • Manufactured from 3.3 expansion borosilicate glass complying with ISO 3585 (ASTM E-438 Type 1 Class A)
  • For use with 47mm diameter membranes
  • Complete assembly comprises of four components: - 5809/2 Graduated 300 ml capacity funnel - 5810/3 Support base with porosity 3 sintered filter disc and vacuum connection with 40/38 ground socket joint - 5810/4 1000 ml conical flask suitable for use under vacuum with 40/38 ground cone joint - 5810/5 Metal anodised retaining clamp
  • Fully autoclavableIdeal for microbiology and preparation of HPLC solvents
  • Support base has plastic screw thread tubing connector to accept 9mm bore vacuum tubing
  • Each component is also available to order separately
Información legal
PYREX is a registered trademark of Corning, Inc.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)