Merck
S64239
N-(2,2,2-TRICHLORO-1-((MORPHOLINE-4-CARBOTHIOYL)-AMINO)-ETHYL)-BENZAMIDE
AldrichCPR
Seleccione un Tamaño
25MG
29,10 €
29,10 €
Fecha estimada de envío13 de febrero de 2025Detalles
Seleccione un Tamaño
Cambiar Vistas
25MG
29,10 €
About This Item
Fórmula lineal:
C14H16Cl3N3O2S
Número de CAS:
Peso molecular:
396.726
Número MDL:
29,10 €
Fecha estimada de envío13 de febrero de 2025Detalles
Otras notas
Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of rare and unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.
NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH'S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT AS-IS AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)