Documentos clave
PL800GA-NIT-8
Plas-Labs replacement nitrile gloves
for 2 piece glove set, size 8 (for 8 in. ports)
Seleccione un Tamaño
1 PAIR
114,00 €
114,00 €
Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad
Seleccione un Tamaño
Cambiar Vistas
1 PAIR
114,00 €
About This Item
Código UNSPSC:
46181504
NACRES:
NB.35
114,00 €
Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad
Materiales
nitrile
tamaño
8 (for 8 in. ports)
Características
reusable
for use with glove box sleeves
envase
pack of 1 pair
fabricante / nombre comercial
Plas-Labs 800GA-NIT-8
Descripción general
Plas-Labs glovebox gloves are available as either one-piece or two-piece assemblies with interchangeable sleeves and gloves. This enables personnel to use the same glove box by changing the size or type of glove being used. Choose from four rubber types. Gloves are 15 mil thickness unless noted otherwise.
Note: Order sleeves, gloves, and glove over sleeve connectors separately.
  • White Hypalon® rubber for resistance to abrasion, oxidizing chemicals, ozone and UV light
  • Black neoprene rubber for chemical resistance
  • Black butyl rubber for highest impermeability to water vapor, gases, and toxic fluids
  • Green nitrile rubber for resistance to oil, fuel, and acids
  • Latex rubber for flexibility
Información legal
Hypalon is a registered trademark of E. I. du Pont de Nemours and Company
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)