Merck
PH005229
Sigma-Aldrich
2-Benzyl-9H-fluoreno[3,2-d][1,3]oxazole
AldrichCPR
About This Item
Fórmula lineal:
C21H15NO
Número de CAS:
cadena SMILES
C12=CC(N=C3CC4=CC=CC=C4)=C(O3)C=C1CC5=CC=CC=C25
InChI
1S/C21H15NO/c1-2-6-14(7-3-1)10-21-22-19-13-18-16(12-20(19)23-21)11-15-8-4-5-9-17(15)18/h1-9,12-13H,10-11H2
Otras notas
Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.
NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.
Punto de inflamabilidad (°F)
Not applicable
Punto de inflamabilidad (°C)
Not applicable
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)