Documentos clave
CAD7977
Micro-emulsifying needle
with reinforcing bar, gauge 18 ×, L 2 7/8 in.
Seleccione un Tamaño
4 EA
645,00 €
645,00 €
Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad
Seleccione un Tamaño
Cambiar Vistas
4 EA
645,00 €
About This Item
Código UNSPSC:
42142546
NACRES:
NB.03
645,00 €
Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad
envase
pack of 4 ea
fabricante / nombre comercial
Cadence Science 7977
Longitud
2 7/8 in.
manómetro
18 ×
Descripción general
Widely used for allergens and the mixing of adjuvants and antigens, oil based liquids or any two liquid media differing in viscosity. By locking two syringes on to the double hub needle or double hub adapter, two liquids such as a mineral oil and an emulsifying agent may be readily pumped between the two syringes to produce a uniform mixture.
Indicated lengths are the overall length. Researchers and technicians utilize Micro-Mate® 5 mL and 10 mL Luer lock tip glass syringes for micro-emulsifying procedures. The outstanding quality of the glass and the special secure Luer locking tip provide quality and reliability.
Información legal
Micro-Mate is a registered trademark of Cadence Science
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)