Documentos clave
CAD6311
1-way threaded end adapter (UTS)
MLL to 10-32 internal standard thread (plated brass)
Seleccione un Tamaño
10 EA
390,00 €
390,00 €
Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad
Seleccione un Tamaño
Cambiar Vistas
10 EA
390,00 €
About This Item
Código UNSPSC:
41121815
NACRES:
NB.03
390,00 €
Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad
envase
pack of 10 ea
fabricante / nombre comercial
Cadence Science 6311
Descripción general
  • Plated brass or 316 stainless steel
  • Meets ISO 594 specifications
  • Final assembly water tested at 45 PSI
Female Luer Lock (FLL) - contains a tabbed hub for secure fitting to a threaded male Luer lock.
Micro-Mate® Female Luer Lock (MMFLL) - designed and manufactured to reduce the stripping of threads which occurs when metal adapters are mated with plastic male Luer lock adapters. Uniquely designed to facilitate easy cleaning and maintenance.
Male Luer Lock (MLL) - male Luer slip with a threaded sheath for a secure fitting to a female Luer lock.
Male Luer (ML) - male Luer slip that fits with a female Luer.
Información legal
Micro-Mate is a registered trademark of Cadence Science
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)