Documentos clave
BR718920
BRAND® counting chamber BLAUBRAND® Bürker pattern
with clips, double ruled
Sinónimos:
BLAUBRAND®, Bürker, Cell culture, Counting Chambers, ldpe bottle, polyethylene bottles, polyethylene containers
Seleccione un Tamaño
1 EA
83,30 €
83,30 €
Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad
Seleccione un Tamaño
Cambiar Vistas
1 EA
83,30 €
About This Item
Código UNSPSC:
41122600
NACRES:
NB.22
83,30 €
Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad
Materiales
colorless
glass
esterilidad
non-sterile
Características
with clips
fabricante / nombre comercial
BRAND 718920
pocillos
1
Descripción general
Manufactured according to DIN 12 847, including 2 haemacytometer cover glasses.The ruling shows 9 large squares of 1 mm² each. These are used for counting leucocytes.Each large square is subdivided by double lines (0.05 mm apart) into 16 group squares with 0.2 mm sides. The group squares correspond in size to the Neubauer counting chamber, but have no further subdivisions. They are used for counting thrombocytes and erythrocytes. The double lines from mini squares with an area of 0.0025 mm².
Características y beneficios
• BLAUBRAND® quality• Outstanding measurement precision• 100% certified quality
Envase
1 counting chamber, complete with 2 haemacytometer cover glasses, in transparent plastic box.
Información legal
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)