Documentos clave
BAH182140000
SP Bel-Art® Plastic Spring Style Tubing Clamps
for tubing o.d., up to 5/32 in. (12 mm)
About This Item
Código UNSPSC:
41122800
NACRES:
NB.43
Características
for tubing o.d. up to 5/32 in. (12 mm)
envase
pack of 3 ea
fabricante / nombre comercial
Bel-Art H18214-0000
Descripción general
Perfect for all flexible tubing, this positive-action, self-closing tube clamp opens by squeezing the handles together.
Características y beneficios
  • Chemical and stain-resistant
  • Made of Acetal plastic and stainless steel
Información legal
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)