Documentos clave
BAF420751002
SP Bel-Art® Secador® Mini Gas-Purge Desiccator Cabinet
polystyrene, volume 0.3 ft3
Seleccione un Tamaño
1 EACH
376,00 €
376,00 €
Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad
Seleccione un Tamaño
Cambiar Vistas
1 EACH
376,00 €
About This Item
Código UNSPSC:
41123002
NACRES:
NB.77
376,00 €
Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad
Materiales
clear
polystyrene
envase
pkg of 1 ea
fabricante / nombre comercial
Bel-Art F42075-1002
volumen
0.3 ft3
Descripción general
The sturdy construction and compact size of this desiccator cabinet assures reliable storage for moisture sensitive products when benchtop space is at a premium.
  • Large side swinging door maximizes access to interior space; has a patented seal design with tab latches and security loop for padlock or tamper-evident seals; opens right to left
  • Top mounted gas ports are tapered to accept tubing with ¼ to ⅜” I.D.
  • Door mounted dial hygrometer indicates relative humidity at a glance
  • Clear polystyrene for easy viewing of contents
  • Gas ports can be closed to use desiccator with standard desiccant cartridges
Información legal
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Secador is a registered trademark of SP Scienceware, a division of SP
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)