Skip to Content
Merck

Skip To

V800293

Perchloric acid

AR, ≥70%

Synonym(s):

PCA

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
HClO4
CAS Number:
Molecular Weight:
100.46
UNSPSC Code:
12352300
PubChem Substance ID:
EC Number:
231-512-4
MDL number:
Concentration:
≥70%
Grade:
AR
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

AR

product line

Vetec

concentration

≥70%

SMILES string

OCl(=O)(=O)=O

InChI

1S/ClHO4/c2-1(3,4)5/h(H,2,3,4,5)

InChI key

VLTRZXGMWDSKGL-UHFFFAOYSA-N

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Perchloric acid


signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Ox. Liq. 1 - Skin Corr. 1A - STOT RE 2

target_organs

Thyroid

Storage Class

5.1A - Strongly oxidizing hazardous materials

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



John Greenwood et al.
ACS nano, 9(5), 5520-5535 (2015-04-22)
We shine light on the covalent modification of graphite and graphene substrates using diazonium chemistry under ambient conditions. We report on the nature of the chemical modification of these graphitic substrates, the relation between molecular structure and film morphology, and
H Harrington Cleveland et al.
Development and psychopathology, 27(1), 51-67 (2015-02-03)
Data drawn from the in-home subsample of the PROSPER intervention dissemination trial were used to investigate the moderation of intervention effects on underage alcohol use by maternal involvement and candidate genes. The primary gene examined was dopamine receptor D4 (DRD4).
Dipanwita Das et al.
Journal of the American Chemical Society, 135(10), 4018-4026 (2013-02-28)
Catalytic four-electron reduction of O2 by ferrocene (Fc) and 1,1'-dimethylferrocene (Me2Fc) occurs efficiently with a dinuclear copper(II) complex [Cu(II)2(XYLO)(OH)](2+) (1), where XYLO is a m-xylene-linked bis[(2-(2-pyridyl)ethyl)amine] dinucleating ligand with copper-bridging phenolate moiety], in the presence of perchloric acid (HClO4) in



Questions

Reviews

No rating value

Active Filters