Skip to Content
Merck

Skip To

1044265

USP

Atenolol Related Compound B

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

2-[4-(2,3-Dihydroxypropoxy)phenyl]acetamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
25 mg
Please contact Customer Service for Availability
€1,160.00

About This Item

Empirical Formula (Hill Notation):
C11H15NO4
CAS Number:
Molecular Weight:
225.24
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€1,160.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

atenolol

packaging

pkg of 25 mg

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecules)

format

neat

storage temp.

2-8°C

SMILES string

NC(=O)Cc1ccc(cc1)OCC(O)CO

InChI

1S/C11H15NO4/c12-11(15)5-8-1-3-10(4-2-8)16-7-9(14)6-13/h1-4,9,13-14H,5-7H2,(H2,12,15)

InChI key

CQOQCZLTCMUVMX-UHFFFAOYSA-N

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Application

Atenolol Related Compound B USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Atenolol
  • Atenolol Tablets

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
104425410442761044298
application(s)

pharmaceutical (small molecules)

application(s)

pharmaceutical (small molecules)

application(s)

pharmaceutical (small molecules)

application(s)

pharmaceutical (small molecules)

format

neat

format

neat

format

neat

format

neat

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

packaging

pkg of 25 mg

packaging

pkg of 50 mg

packaging

pkg of 25 mg

packaging

pkg of 25 mg

API family

atenolol

API family

atenolol

API family

atenolol

API family

atenolol


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters