Skip to Content
Merck
All Photos(1)

Documents

PHL89744

Oleandrin

phyproof® Reference Substance

Synonym(s):

5β,20(22)-Cardenolide-3β,14,16β-triol-3-([2,6-dideoxy-3-O-methyl-α-L-arabinohexopyranosyl]oxy) 16-acetate, Corrigen, Foliandrin, Folinerin, Neriol, Neriolin, Neriostene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H48O9
CAS Number:
Molecular Weight:
576.72
UNSPSC Code:
41116107
NACRES:
NA.24

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥95.0% (HPLC)

manufacturer/tradename

PhytoLab

application(s)

food and beverages

format

neat

SMILES string

CO[C@H]1C[C@@H](O[C@@H](C)[C@@H]1O)O[C@H]2CC[C@@]3(C)C(CCC4C3CC[C@]5(C)[C@H]([C@H](C[C@]45O)OC(C)=O)C6=CC(=O)OC6)C2

InChI

1S/C32H48O9/c1-17-29(35)24(37-5)14-27(39-17)41-21-8-10-30(3)20(13-21)6-7-23-22(30)9-11-31(4)28(19-12-26(34)38-16-19)25(40-18(2)33)15-32(23,31)36/h12,17,20-25,27-29,35-36H,6-11,13-16H2,1-5H3/t17-,20+,21-,22-,23+,24-,25-,27-,28-,29-,30-,31+,32-/m0/s1

InChI key

JLPDBLFIVFSOCC-XYXFTTADSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Application

  • Cardiac glycoside-induced anti-cancer effects of oleandrin: Research highlighted oleandrin′s role in the negative regulation of thyroid adenoma-associated protein (THADA), contributing to its potential as an anti-cancer agent in cardiac glycoside studies (Katoh et al., 2024).
  • Oleandrin in veterinary medicine: A study investigated the safe oral dosage of oleandrin for Beagles, providing crucial data on its pharmacological safety in veterinary applications (Fossum et al., 2024).

Biochem/physiol Actions

Cardiac glycoside, inhibits Na(+)/K(+) ATPase; potent carcinostatic cardiac glycoside.

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service