Skip to Content
Merck
All Photos(1)

Key Documents

822794

TSKgel® SuperMP(PW)-M (8 µm) HPLC Columns

L x I.D. 3.5 cm × 4.6 mm, hydroxylated methacrylate phase

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€809.00

€809.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EA
€809.00

About This Item

UNSPSC Code:
41115711

€809.00


Please contact Customer Service for Availability

Product Name

TSKgel® SuperMP(PW)-M Guard Column, phase hydroxylated methacrylate, L × I.D. 3.5 cm × 4.6 mm, 8 μm particle size

material

stainless steel

Agency

suitable for USP L38
suitable for USP L39

product line

TSKgel®

manufacturer/tradename

Tosoh 22794

parameter

≤0.5 M salt concentration
≤20% polar organic solvent
0.6 mL/min flow rate
10-80 °C temperature

technique(s)

HPLC: suitable

L × I.D.

3.5 cm × 4.6 mm

matrix

hydroxylated methacrylic polymer

matrix active group

hydroxylated methacrylate phase

particle size

8 μm

operating pH

2-12

separation technique

size exclusion (SEC)

General description

The innovative multi-pore particle synthesis technology which Tosoh scientists pioneered, has now been extended to TSKgel® SuperMultiporePW columns and guard columns for water-soluble polymer analysis. Three semi-micro columns varying in linear range are available within this series, enabling high speed and high resolution analysis with lowered solvent consumption. The base material of TSKgel SuperMultiporePW columns and guard columns is polymethacrylate.A wide molecular mass range can be analyzed with the three different TSKgel SuperMultiporePW columns, from high molecular mass water-soluble polymers to oligomers. The packing material in the TSKgel SuperMultiporePW columns is more hydrophilic than that of TSKgel PWxl series columns, which further reduces the chance of adsorption of hydrophilic polymers.

Physical form

Shipped in water.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service