2630905
Inlet Liner, Splitless Type, Straight Design (unpacked)
pkg of 5 ea
All Photos(1)
About This Item
UNSPSC Code:
12352200
Recommended Products
packaging
pkg of 5 ea
L × O.D. × I.D.
92 mm × 6.35 mm × 4.0 mm
General description
Deactivated (silanized) glass inlet liner for capillary injection ports.
- Fits PerkinElmer (AutoSystem)
Choose from one of the most recent versions:
Certificates of Analysis (COA)