Key Documents
21911
SGE Syringes for Agilent/HP 7673, 7683 and 6850 ALS Autosampler, Fixed Needle
volume 5 μL, SK-5F-HP-0.63, needle size 23 ga, needle L 42 mm, pkg of 6 ea
Select a Size
6 EA
€546.00
€546.00
Please contact Customer Service for Availability
Request a Bulk Order
Select a Size
Change View
6 EA
€546.00
About This Item
UNSPSC Code:
41122001
€546.00
Please contact Customer Service for Availability
Request a Bulk Order
material
cone tip stainless steel needle (point style 1)
needle size
23 ga
feature
needle type F (cemented needle)
packaging
pkg of 6 ea
manufacturer/tradename
SGE 001814
needle L
42 mm
volume
5 μL
compatibility
for use with Agilent/HP 7673 and 7683 autosampler
General description
SGE syringes are designed and tested to meet critical autosampler specifications. These syringes are designed with a unique plunger to eliminate bending and seizing. The needle point style withstands multiple and fast septum injections, making it one of the most troublefree autosampler syringes available.
Choose from one of the most recent versions:
Certificates of Analysis (COA)