Key Documents
26889-U
SGE Syringes for Agilent/HP 7673, 7683 and 6850 ALS Autosampler, Fixed Needle
volume 10 μL, 10F-HP-0.63/0.47, needle size 23-26 ga, needle L 42 mm
Select a Size
6 EA
€381.00
€381.00
Please contact Customer Service for Availability
Request a Bulk Order
Select a Size
Change View
6 EA
€381.00
About This Item
UNSPSC Code:
41122001
NACRES:
NB.03
€381.00
Please contact Customer Service for Availability
Request a Bulk Order
material
cone tip stainless steel needle
needle size
23-26 ga
feature
needle type F (cemented needle)
point style #1
packaging
pkg of 6 ea
manufacturer/tradename
SGE 002822
needle L
42 mm
volume
10 μL
General description
SGE syringes are designed and tested to meet critical autosampler specifications. These syringes are designed with a unique plunger to eliminate bending and seizing. The needle point style withstands multiple and fast septum injections, making it one of the most troublefree autosampler syringes available.
Choose from one of the most recent versions:
Certificates of Analysis (COA)